Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEFTY2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542105
Description
Peptide sequence: MWPLWLCWAL WVLPLAGPGA ALTEEQLLGS LLRQLQLSEV PVLDRADMEK Sequence homology: Cow: 79%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 86%; Rat: 83%.
Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
Specifications
| LEFTY2 | |
| Polyclonal | |
| Unconjugated | |
| LEFTY2 | |
| 6030463A22Rik; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; LEFTY2; LEFTYA; MGC46222; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7044 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00292 | |
| LEFTY2 | |
| Synthetic peptide directed towards the N-terminal of human LEFTY2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction