Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEFTY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109905 Supplier Invitrogen™ Supplier No. PA5109905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRAQYVVLLR RSHGDRSRGK RFSQSFREVA GRFLASEAST HLLVFGMEQR LPPNSELVQA VLRLFQEPVP KAALHRHGRL SPRSAQARVT VEWLRVRDDG SNRTSLIDSR LVSVHESGWK AFDVTEAVNF WQQLSRPRQP LLLQVSVQRE HLGPLASGAH KLVRFASQGA PAGLGEPQLE LHTLDLRDYG AQGDCDPEAP MTEGTRCCRQ EMYIDLQGMK WAKNWVLEPP GFLAYECVGT CQQPPEALAF NWPFLGPRQC IASETASLPM IVSIKEGGRT RPQVVSLPNM RVQKCSCASD GALVPRRLQP.

Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEFTY2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LEFTY2
Gene Accession No. O00292, P57785
Gene Alias 6030463A22Rik; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; LEFTY2; LEFTYA; MGC46222; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4
Gene Symbols LEFTY2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 57-366 of human LEFTY2 (NP_003231.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320202, 7044
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.