Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legionella Pneumophila Antibody (2F10), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102520AF405
Description
Legionella Pneumophila Monoclonal specifically detects Legionella Pneumophila in Bacteria samples. It is validated for ELISA, Sandwich ELISA.Specifications
Legionella Pneumophila | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with membrane fraction of Legionella pneumophila, strain Philadelphia 1. | |
0.2 mL | |
Primary | |
Bacteria | |
Purified |
ELISA | |
2F10 | |
ELISA, Sandwich ELISA | |
Mouse | |
Protein G purified | |
RUO | |
Reacts with LPS membrane of Legionella pneumophila. No cross-reactivity with Legionella pneumophila serogroups 2, 6, 7, 10 and 11, Legionella bozemanii, Legionella lonbeachae, Legionella micdadei, Corynebacterium diphtheria, Pseudomonas fluorescens, Pseudomonas cepacia, Pseudomonas aerugenose, Bordetella pertusis, Leptospira interrogens (Australia, Pomona, Icterogemorrhagia), Toxoplasma gondii, Hemophilus influenza (type B), Hemophilus ducreyi, Brucella abortus, Bacillus anthracis, Klebsiella pneumoniae, Listeria monocytogenes, Mycobacterium tuberculosis, Neisseria meningitides, Pasteurella multocida, Proteus vulgaris, Shigella sonnei, Staphylococcus faecalis, Streptococcus puogenes, Streptococcus pneumoniae, Yersinia pseudotuberculosis, Yersinia pestis EV76, Salmonella typhi, Escherichia coli K88 and Francisella tularensis 15. Cross-reactivity with Legionella pneumophila serogroups 3 and 4 has not been tested. | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction