Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legionella Pneumophila Antibody (LP101), Alexa Fluor™ 647

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064476AF647
Description
Legionella Pneumophila Monoclonal specifically detects Legionella Pneumophila in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Legionella Pneumophila | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Legionella pneumophila serogroups 4,10,14. | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Immunocytochemistry, Immunofluorescence | |
LP101 | |
Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
RUO | |
NB100-64476 is specific for Legionalla pneumophila SS4 (Los Angeles 1). This antibody also has some minor reactivity with SG14 (1586-SCT-H) and SG10 (Leiden). There is no cross reactivity with SG1, 3, 5, 6, 7, 9, 11, 12 or 13, Listeria erythra, Listeria dumoffii, Escherichia coli or Pseudomonas aeruginosa. | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction