Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Legionella pneumophila LPS Monoclonal Antibody (F93D)
GREENER_CHOICE

Catalog No. PIMA518214
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518214 Supplier Invitrogen™ Supplier No. MA518214
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is reactive with LPS of all L. pneumophila strains tested (11). Purity is >90%. Recommended pairs for ELISA: MA5-18213 (capture) and MA5-18214 (conjugated, detection).

The causative agent of what is commonly called Legionnaire's Disease, Legionella pneumophila causes acute pneumonia. The disease can also present itself as an nonpneumonic, influenza-like illness called Pontiac Fever. Legionella is found in aquatic environments where it's growth is supported by amoebae. In human infections Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Legionella pneumophila LPS
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone F93D
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias L. pneumophila; Legionellosis
Host Species Mouse
Immunogen Legionella pneumophila native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.