Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Legionella pneumophila Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA173142
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173142 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173142 Supplier Invitrogen™ Supplier No. PA173142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73142 antibody reacts with Legionella pneumophila. PA1-73142 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73142 antibody was raised against whole cell preparation of Legionella pneumophila; ATCC #33152.

The causative agent of what is commonly called Legionnaire's Disease, Legionella pneumophila causes acute pneumonia. The disease can also present itself as an nonpneumonic, influenza-like illness called Pontiac Fever. Legionella is found in aquatic environments where it's growth is supported by amoebae. In human infections Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Legionella pneumophila
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 4-5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias L. pneumophila; Legionellosis
Host Species Rabbit
Immunogen Whole cell preparation of Legionella pneumophila; ATCC #33152.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.