Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Legionella pneumophila Serogroup 1 Monoclonal Antibody (142)
GREENER_CHOICE

Catalog No. PIMA546573
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546573 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546573 Supplier Invitrogen™ Supplier No. MA546573
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Legionella pneumophila Serogroup 1 Monoclonal Antibody for Western Blot, ICC/IF, IP

Legionella pneumophila serogroup 1 is the most virulent strain causing the majority of infections. The remaining non-pneumophila species (found in water and soil) are considered nonpathogenic until shown to cause disease. Of the CDC reported cases less than 5% is attributed to non-pathognenic species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Legionella pneumophila Serogroup 1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 142
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias L. pneumophila
Host Species Mouse
Immunogen Native Legionella pneumophila.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.