Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Legionella pneumophila Serogroup 1 Recombinant Rabbit Monoclonal Antibody (6026)
GREENER_CHOICE

Catalog No. PIMA533306
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533306 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533306 Supplier Invitrogen™ Supplier No. MA533306
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specific to all serogroup 1 strains tested (8). Negative for human cells.

Legionella pneumophila serogroup 1 is the most virulent strain causing the majority of infections. The remaining non-pneumophila species (found in water and soil) are considered nonpathogenic until shown to cause disease. Of the CDC reported cases less than 5% is attributed to non-pathognenic species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Legionella pneumophila Serogroup 1
Applications ELISA, Lateral Flow, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6026
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias L. pneumophila
Host Species Rabbit
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.