Learn More
Invitrogen™ Legionella pneumophila subsp. pneumophila (strain Philadelphia 1/ATCC 33152/DSM 7513) sdeA Polyclonal Antibody

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Legionella pneumophila subsp. pneumophila (strain Philadelphia 1/ATCC 33152/DSM 7513) sdeA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.88 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Host Species | Rabbit |
| Immunogen | Recombinant Legionella pneumophila subsp Ubiquitinating/deubiquitinating enzyme SdeA protein (19-268aa). |
| Purification Method | Protein G |
| Quantity | 100 μL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.