Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legumain/Asparaginyl Endopeptidase Antibody (46I3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | Legumain/Asparaginyl Endopeptidase |
|---|---|
| Clone | 46I3 |
| Concentration | LYOPH |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA.Specifications
| Legumain/Asparaginyl Endopeptidase | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| LGMN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 46I3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Q99538 | |
| 5641 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title