Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ LEI 101 hydrochloride
GREENER_CHOICESDP

Catalog No. 582610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-261-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 58-261-0 Supplier R&D Systems™ Supplier No. 5826/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective CB2 partial agonist; orally biovailable

Potent and selective CB2 partial agonist (pEC50 = 8.0). Exhibits >100-fold selectivity for CB2 over CB1 and no significant interaction with hFAAH, MAGL, DAGL or NAPE-PLD. Displays antinociceptive activity in a rat neuropathic pain model and attenuates renal inflammation in a mouse kidney damage model. Orally bioavailable with low brain penetration.

Chemical Identifiers

CAS 2250025-91-1

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Formula Weight 508.99
Physical Form Solid
Purity >98%
Chemical Name or Material 3-Cyclopropyl-1-[[4-[6-[(1,1-dioxido-4-thiomorpholinyl)methyl]-5-fluoro-2-pyridinyl]phenyl]methyl]-2
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.