Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Leishmania gp63 Monoclonal Antibody (96-126)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA181830
Description
MA1-81830 recognizes living and fixed parasites, but will not bind to denatured or improperly folded recombinant gp63. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
The causative agent of what is commonly called Legionnaire's Disease, Legionella pneumophila causes acute pneumonia. The disease can also present itself as an nonpneumonic, influenza-like illness called Pontiac Fever. Legionella is found in aquatic environments where it's growth is supported by amoebae. In human infections Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.
Specifications
Leishmania gp63 | |
Monoclonal | |
Conc. Not Determined | |
ascites with no preservative | |
Mouse | |
500 μL | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Immunocytochemistry | |
96-126 | |
Unconjugated | |
L. pneumophila; Legionellosis | |
Recombinant baculovirus expressed in L. major gp63. | |
RUO | |
Protozoa | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction