Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Leishmania LPG Monoclonal Antibody (CA7AE)
GREENER_CHOICE

Catalog No. PIMA183817
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183817 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183817 Supplier Invitrogen™ Supplier No. MA183817
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 0.5 mL of distilled water. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles. Mouse anti Leishmania lipophosphoglycan antibody, clone CA7AE recognizes lipophosphoglycan (LPG) the major cell surface glycoconjugate of Leishmania parasites.

Lipophosphoglycan (LPG) is the major cell surface glycoconjugate of Leishmania parasites. LPG antibodies recognize the repeat carbohydrate epitope of most species of Leishmania. The epitope is also found on the excreted acid phosphatase of Leishmania and is expressed on the surface of Leishmania infected macrophages (Tolson et al. 1990).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leishmania LPG
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CA7AE
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with no preservative
Gene Alias L. pneumophila; Legionellosis
Host Species Mouse
Immunogen Heat killed L. donovani promastigotes.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4°C
Product Type Antibody
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.