Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEKR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139808
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MDHHIPMHAL PEEIQKMLPE EKVCKYCGVS YLILHEFKAM EEKVKAMEKE MKFYQGSVDR EKRLQEKLHS LSQELEQYKI DNKSKTERIY DVG.
LEKR1 is a protein coding gene.
Specifications
| LEKR1 | |
| Polyclonal | |
| Unconjugated | |
| LEKR1 | |
| EG546798; EG624866; Gm6534; LEKR1; leucine, glutamate and lysine rich 1; leucine-, glutamate- and lysine-rich protein 1; RGD1562738; RGD1565425 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 389170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZMV7 | |
| LEKR1 | |
| Recombinant protein corresponding to Human LEKR1. Recombinant protein control fragment (Product #RP-108861). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction