Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEKR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560598
Description
Immunogen sequence: KLHKSHIRYT EESNSKEKEI ENLKNLVAEF ESRLKKEIDS NDSVSENLRK EMEQKSDELK RVMLAQTQLI EQFNQSQEEN TFLQETVRRE CEERFEL Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 78%.
LEKR1 is a protein coding gene.
Specifications
LEKR1 | |
Polyclonal | |
Unconjugated | |
LEKR1 | |
EG546798; EG624866; Gm6534; LEKR1; leucine, glutamate and lysine rich 1; leucine-, glutamate- and lysine-rich protein 1; RGD1562738; RGD1565425 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMV7 | |
LEKR1 | |
Recombinant protein corresponding to Human LEKR1. Recombinant protein control fragment (Product #RP-100290). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction