Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LELP1 Antibody (OTI3D5), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272412AF532
Description
LELP1 Monoclonal specifically detects LELP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LELP1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
LELP1 | |
Full length recombinant protein of human LELP1 (NP_001010857) produced in E.coli. | |
0.1 mL | |
149018.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3D5 | |
Western Blot, Immunohistochemistry | |
late cornified envelope-like proline-rich 1, novel small proline-rich protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction