Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEM3 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | LEM3 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LEM3 Polyclonal specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
LEM3 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3 | |
ANKLE1 | |
IgY | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Chicken | |
Protein Kinase | |
PBS with 0.02% Sodium Azide | |
126549 | |
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL | |
Primary | |
Two isoforms of ANKLE1 are known to exist. ANKLE1 antibody is predicted to not cross-react with ANKLE2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title