Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEM3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241144MFV450
Description
LEM3 Polyclonal antibody specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| LEM3 | |
| Polyclonal | |
| Western Blot, ELISA | |
| ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL | |
| 0.1 mL | |
| Protein Kinase | |
| 126549 | |
| Store at 4C in the dark. | |
| IgY |
| Western Blot, ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction