Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572733
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572733 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572733 Supplier Invitrogen™ Supplier No. PA572733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: At least six isoforms of LEMD1 are known to exist; this antibody will detect the longest isoform. LEMD1 antibody is predicted to not cross-react with LEMD2 and LEMD3.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LEMD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LEMD1
Gene Accession No. Q14C37, Q68G75
Gene Alias 4930540I23Rik; 6430514M23Rik; AI428128; Cancer/testis antigen 50; CT50; LEM domain containing 1; LEM domain protein 1; LEM domain-containing protein 1; Lemd1; LEMP-1
Gene Symbols LEMD1
Host Species Rabbit
Immunogen LEMD1 antibody for a 19 amino acid synthetic peptide near the center of human LEMD1. The immunogen is within amino acids 90 - 140 of LEMD1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213409, 93273
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.