Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEMD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598781
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598781 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598781 Supplier Invitrogen™ Supplier No. PA598781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LEMD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The LEM (lamina-associated polypeptide-emerin-MAN1) domain is a motif shared by a group of lamin-interacting proteins of the inner nuclear membrane (INM) and nucleoplasm. One such protein, LEMD2 (LEM domain-containing protein 2), is a lamina-associated protein in the INM involved in nuclear structure organization, containing an N-terminal LEM motif, two transmembrane domains, and a MAN1-Src1p C-terminal (MSC) domain. LEMD2 is a truncated paralog of MAN1, lacking the MAN1-specific C-terminal RNA-recognition motif, but like MAN1 is required for myogenic differentiation. Mutations in LEMD2 are thought be among the possible causes of muscle disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEMD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.475 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LEMD2
Gene Accession No. Q8NC56
Gene Alias BC026588; CTRCT42; dJ482C21.1; hLEM2; LEM domain containing 2; LEM domain-containing protein 2; Lem2; LEMD2; NET25; nuclear envelope constituent; Nuclear envelope transmembrane protein 25
Gene Symbols LEMD2
Host Species Rabbit
Immunogen Recombinant Human LEM domain-containing protein 2 protein (17-204AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221496
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.