Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lenalidomide
GREENER_CHOICESDP

Catalog No. 6305100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-051-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 63-051-00 Supplier R&D Systems™ Supplier No. 6305/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cereblon binder; induces ubiquitination and degradation of CK1α by E3 ubiquitin ligase

Thalidomide analog. Immune modulatory drug and cereblon binding compound. Induces ubiquitination and degradation of casein kinase (CK) 1α by the E3 ubiquitin ligase CRL4CRBN. Also TNF-α inhibitor and angiogenesis inhibitor. Promotes degradation of transcription factor SALL4.

Chemical Identifiers

CAS 191732-72-6
Molecular Formula C13H13N3O3

Specifications

Quantity 100 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Formula Weight 259.26
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 259.265
Chemical Name or Material 3-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.