Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LENG1 Antibody (OTI10A7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272202APC
Description
LENG1 Monoclonal specifically detects LENG1 in Human samples. It is validated for Flow Cytometry.Specifications
| LENG1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| LENG1 | |
| Full length recombinant protein of human LENG1 (NP_077292) produced in HEK293T cells. | |
| 0.1 mL | |
| 79165.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI10A7 | |
| Flow Cytometry | |
| leukocyte receptor cluster (LRC) member 1, leukocyte receptor cluster member 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction