Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LENG1 Antibody (OTI10A7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272202PE
Description
LENG1 Monoclonal specifically detects LENG1 in Human samples. It is validated for Flow Cytometry.Specifications
LENG1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
LENG1 | |
Full length recombinant protein of human LENG1 (NP_077292) produced in HEK293T cells. | |
0.1 mL | |
79165.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI10A7 | |
Flow Cytometry | |
leukocyte receptor cluster (LRC) member 1, leukocyte receptor cluster member 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction