Learn More
LENG1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304714A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function of this protein remains unknown.Specifications
LENG1 | |
Polyclonal | |
Unconjugated | |
LENG1 | |
leukocyte receptor cluster (LRC) member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79165 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96BZ8 | |
LENG1 | |
Between 200 and 250 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.