Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LENG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100602 Supplier Invitrogen™ Supplier No. PA5100602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LENG8.

LENG8 (Leukocyte Receptor Cluster Member 8) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LENG8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LENG8
Gene Accession No. Q8CBY3, Q96PV6
Gene Alias AW049671; KIAA1932; LENG8; leukocyte receptor cluster (LRC) member 8; leukocyte receptor cluster member 8; Leukocyte receptor cluster member 8 homolog; MGC40108; pp13842
Gene Symbols LENG8
Host Species Rabbit
Immunogen A synthesized peptide derived from human LENG8(Accession Q96PV6), corresponding to amino acid residues H431-K481.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114823, 232798, 361506
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.