Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LENG8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559847
Description
Immunogen sequence: PERELKKQKR AARFQHGHSR RLRLEPLVLQ MSSLESSGAD PDWQELQIVG TCPDITKHYL RLTCAPDPST VRPVAVLKKS LCMVKCHWKE KQDYAFACEQ MKSIRQDLTV Q Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
LENG8 (Leukocyte Receptor Cluster Member 8) is a Protein Coding gene.
Specifications
LENG8 | |
Polyclonal | |
Unconjugated | |
LENG8 | |
AW049671; KIAA1932; LENG8; leukocyte receptor cluster (LRC) member 8; leukocyte receptor cluster member 8; Leukocyte receptor cluster member 8 homolog; MGC40108; pp13842 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PV6 | |
LENG8 | |
Recombinant protein corresponding to Human LENG8. Recombinant protein control fragment (Product #RP-99146). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction