Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT

Catalog No. p-7182875
Encompass_Preferred
Click to view available options
Quantity:
100 μL
1 mL
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.

Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.

The Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice.

Recommended Storage

Store at -68 to -85°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 100 μL. Store (at -68°C to -85°C.)
Control Type Positive Control
Delivery Type Lentiviral
Promoter U6
Transfection Technique Lentiviral transduction
Species Human
For Use With (Application) CRISPR gene knockout
Product Line LentiArray™
Product Type CRISPR Positive Control Lentivirus
Quantity 100 μL
Shipping Condition Dry Ice
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.