Learn More
Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.
$634.00 - $2230.00
Specifications
Delivery Type | Lentiviral |
---|---|
Format | Tube |
Control Type | Positive Control |
RNAi Type | gRNA |
Species | Human |
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%.
The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).
Specifications
Lentiviral | |
Positive Control | |
Human | |
LentiArray™ | |
U6 |
Tube | |
gRNA | |
Genome Editing | |
gRNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.