Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEO1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079777IR
Description
LEO1 Polyclonal antibody specifically detects LEO1 in Human, Mouse samples. It is validated for Western BlotSpecifications
LEO1 | |
Polyclonal | |
Western Blot | |
Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), RDLReplicative senescence down-regulated leo1-like protein, RNA polymerase-associated protein LEO1 | |
The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human senescence down regulated leo1-like using the numbering given in TrEMBL entry Q8WVC0 (GeneID 123169). | |
0.1 mL | |
Phospho Specific, Transcription Factors and Regulators | |
123169 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction