Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEO1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219346
Description
LEO1 Polyclonal specifically detects LEO1 in Human samples. It is validated for Western Blot.Specifications
LEO1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), RDLReplicative senescence down-regulated leo1-like protein, RNA polymerase-associated protein LEO1 | |
Rabbit | |
75 kDa | |
0.1 mL | |
Transcription Factors and Regulators | |
123169 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q8WVC0 | |
LEO1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human LEO1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Chimpanzee (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction