Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549415
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549415 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549415 Supplier Invitrogen™ Supplier No. PA549415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LEO1 Polyclonal Antibody for Western Blot

Prostaglandin-endoperoxide synthase (PTGS), also known as cyclooxygenase, is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes the inducible isozyme. It is regulated by specific stimulatory events, suggesting that it is responsible for the prostanoid biosynthesis involved in inflammation and mitogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEO1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Leo1
Gene Accession No. Q5XJE5, Q641X2
Gene Alias Gm185; Leo1; LEO1 homolog, Paf1/RNA polymerase II complex component; Leo1 Paf1/RNA polymerase II complex component; Leo1, Paf1/RNA polymerase II complex component, homolog; Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); RDL; Replicative senescence down-regulated leo1-like protein; RNA polymerase-associated protein LEO1
Gene Symbols Leo1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 616-650 amino acids from the C-terminal region of Mouse Leo1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235497, 300837
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.