Learn More
Leo1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Leo1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2330
![]() |
Bethyl Laboratories, Inc
IHC00573T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2329
![]() |
Bethyl Laboratories, Inc
IHC00573 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla,
The PAF1 complex is a multifunctional complex. The PAF1 complex interacts with POLR2A. It may be involved in both initiation and elongation, histone methylation and RNA processing. Overexpression of LEO1 induces cell growth arrest and premature senescence of fibroblasts.Specifications
Leo1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RDL; RNA polymerase-associated protein LEO1; Replicative senescence down-regulated leo1-like protein | |
Between 550 and 600 | |
Primary | |
4° C | |
LEO1 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q5XJE5, Q8WVC0 | |
123169, 235497 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.