Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEPRE1 Monoclonal Antibody (OTI1H3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526043
Description
LEPRE1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.
Specifications
LEPRE1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32P28 | |
P3H1 | |
Full length human recombinant protein of LEPRE1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H3 | |
Unconjugated | |
P3H1 | |
GROS1; Growth suppressor 1; LEPRE1; leprecan; Leprecan-1; leucine- and proline-enriched proteoglycan 1; leucine proline-enriched proteoglycan (leprecan) 1; OI8; P3H1; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 1; PSEC0109 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64175 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction