Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEPRE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552911 Supplier Invitrogen™ Supplier No. PA552911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVECAKTYLL FFPNDEVMNQ NLAYYAAMLG EEHTRSIGPR ESAKEYRQRS LLEKELLFFA YDVFGIPFVD PDSWTPEEVI PKRLQEKQKS ERETAVRISQ EIGNLMKEIE TLVEEKTKES LDVSRLT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEPRE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P3H1
Gene Accession No. Q32P28
Gene Alias GROS1; Growth suppressor 1; LEPRE1; leprecan; Leprecan-1; leucine- and proline-enriched proteoglycan 1; leucine proline-enriched proteoglycan (leprecan) 1; OI8; P3H1; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 1; PSEC0109
Gene Symbols P3H1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LEPRE1. Recombinant protein control fragment (Product #RP-89689).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.