Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEPREL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553048 Supplier Invitrogen™ Supplier No. PA553048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANPEHMEMQQ NIENYRATAG VEALQLVDRE AKPHMESYNA GVKHYEADDF EMAIRHFEQA LREYFVEDTE CRTLCEGPQR FEEYEYLGYK AGLYEAIADH YMQVLVCQHE CV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEPREL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene P3h2
Gene Accession No. Q8IVL5
Gene Alias 4832416N06; AW553532; leprecan-like 1; leprecan-like protein 1; Leprel1; MCVD; Mlat4; myxoid liposarcoma associated protein 4; myxoid liposarcoma-associated protein 4; P3h2; procollagen-proline 3-dioxygenase 2; prolyl 3-hydroxylase 2; prolyl 3-hydroxylase 3
Gene Symbols P3h2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LEPREL1. Recombinant protein control fragment (Product #RP-89684).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55214
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.