Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEPREL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596918 Supplier Invitrogen™ Supplier No. PA596918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRYGGRQDEN RVPSGVNVEG AEVHGFSMGK KLSPKIDRDL REGGPLLYEN ITFVYNSEQL NGTQRVLLDN VLSEEQCREL HSVASGIMLV GDGYRGKTSP HTPNEKFEGA TVLKALKSGY EGRVPLKSAR LFYDISEKAR RIVESYFMLN STLYFSYTHM VCRTALSGQQ DRRNDLSHPI HADNCLLDPE ANECWKEPPA YTFRDYSALL YMNDDFEGGE FIFTEMDAKT VTASIKPKCG RMISFSSGGE NPHGVKAVTK GKRCAVALWF TLDPLYRELE RIQADEVIAI LDQEQQGKHE L; Positive Samples: NCI-H460; Cellular Location: Endoplasmic reticulum, Golgi apparatus.

This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEPREL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P3h2
Gene Accession No. Q4KLM6, Q8CG71, Q8IVL5
Gene Alias 4832416N06; AW553532; leprecan-like 1; leprecan-like protein 1; Leprel1; MCVD; Mlat4; myxoid liposarcoma associated protein 4; myxoid liposarcoma-associated protein 4; P3h2; procollagen-proline 3-dioxygenase 2; prolyl 3-hydroxylase 2; prolyl 3-hydroxylase 3
Gene Symbols P3h2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human LEPREL1 (NP_0606622).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210530, 288016, 55214
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.