Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEPREL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583676
Description
Immunogen sequence: PMHLQMREDM AKYRRMSGVR PQSFRDLETP PHWAAYDTGL ELLGRQEAGL ALPRLEEALQ GSLAQMESCR ADCEGPEEQQ GAEEEEDGAA SQGGLYEAIA GH.
The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.
Specifications
LEPREL2 | |
Polyclonal | |
Unconjugated | |
P3h3 | |
BC016431; gene rich cluster, B; GRCB; HSU47926; leprecan-like 2; leprecan-like protein 2; LEPREL2; P3H3; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 3; Protein B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10536, 14789, 297595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CG70, Q8IVL6 | |
P3h3 | |
Recombinant protein corresponding to Human P3H3. Recombinant protein control fragment (Product #RP-108184). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction