Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEPREL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596914 Supplier Invitrogen™ Supplier No. PA596914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAAGAGARSG YRGRRSPHTP HERFEGLTVL KAAQLARAGT VGSQGAKLLL EVSERVRTLT QAYFSPERPL HLSFTHLVCR SAIEGEQEQR MDLSHPVHAD NCVLDPDTGE CWREPPAYTY RDYSGLLYLN DDFQGGDLFF TEPNALTVTA RVRPRCGRLV AFSSGVENPH GVWAVTRGRR CALALWHTWA PEHREQEWIE AKELLQESQE EEEEEEEEMP SKDPSPEPPS RRHQRVQDKT GRAPRVREEL; Positive Samples: 22Rv1, NIH/3T3, Mouse skeletal muscle; Cellular Location: Endoplasmic reticulum.

The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEPREL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene P3h3
Gene Accession No. Q8CG70, Q8IVL6
Gene Alias BC016431; gene rich cluster, B; GRCB; HSU47926; leprecan-like 2; leprecan-like protein 2; LEPREL2; P3H3; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 3; Protein B
Gene Symbols P3h3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 487-736 of human LEPREL2 (NP_0550772).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10536, 14789
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.