Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LEPROTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561619 Supplier Invitrogen™ Supplier No. PA561619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRLPFFSKMG TAESEGRETL TQQLPLPAAA MRRLLPASRV STQPVLRLAD SAESLLGRPA LWALGFLLCP PSQAQ Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.

LEPROTL1 negatively regulates growth hormone (GH) receptor cell surface expression in liver. It may play a role in liver resistance to GH during periods of reduced nutrient availability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEPROTL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LEPROTL1
Gene Accession No. O95214
Gene Alias 1110067H13Rik; 1520402O14Rik; AI854296; endospanin-2; HSPC112; LEPROTL1; leptin receptor overlapping transcript like 1; leptin receptor overlapping transcript-like 1; My047; Unknown (protein for MGC:133536); UNQ577/PRO1139; Vps55
Gene Symbols LEPROTL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LEPROTL1. Recombinant protein control fragment (Product #RP-100871).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23484
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.