Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LEPROTL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546034
Description
Peptide sequence: FVLFFYILSP IPYCIARRLV DDTDAMSNAC KELAIFLTTG IVVSAFGLPI Sequence homology: Cow: 100%; Dog: 100%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
LEPROTL1 negatively regulates growth hormone (GH) receptor cell surface expression in liver. It may play a role in liver resistance to GH during periods of reduced nutrient availability.
Specifications
LEPROTL1 | |
Polyclonal | |
Unconjugated | |
LEPROTL1 | |
1110067H13Rik; 1520402O14Rik; AI854296; endospanin-2; HSPC112; LEPROTL1; leptin receptor overlapping transcript like 1; leptin receptor overlapping transcript-like 1; My047; Unknown (protein for MGC:133536); UNQ577/PRO1139; Vps55 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23484 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95214 | |
LEPROTL1 | |
Synthetic peptide directed towards the N-terminal of human LEPROTL1 (aa 37-86). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction