Learn More
Invitrogen™ Leptin Monoclonal Antibody (11)

Description
ABS 022-11-02 detects Leptin in human samples. ABS 022-11-02 has been successfully used in ELISA and Western blot procedures. ABS 022-11-02 binds free Leptin in solution. In sandwich ELISA, ABS 022-11-02 works as a detection antibody with ABS 022-03-02 as a capture antibody. ABS 022-11-02 shows three bands at 16, 32, and 48 kDa in the Western blot application corresponding with the leptin monomer, dimer and trimer forms. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Leptin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 11 |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | Lep |
| Gene Accession No. | P41159 |
| Gene Alias | CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.