Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin/OB Antibody (11), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105047AF350
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for ELISA.Specifications
Leptin/OB | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
LEP | |
Recombinant human leptin adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Primary | |
Clone 11 binds human leptin Cross-reactivity with other species has not been determined | |
Store at 4°C in the dark. |
ELISA | |
11 | |
ELISA | |
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Mouse | |
Protein A or G purified | |
Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism | |
3952.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction