Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin/OB Antibody (6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP221619AF350

Catalog No. NP221619A30


Only null left
Add to Cart

Description

Description

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Leptin/OB
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
LEP
Recombinant human leptin adsorbed onto aluminum hydroxide gel.
0.1 mL
Primary
Human
IgG2b κ
Western Blot, ELISA
6
Western Blot, ELISA
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Mouse
Protein A or G purified
Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism
3952.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.