Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin/OB Antibody (6), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221619AF647
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.Specifications
Leptin/OB | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LEP | |
Recombinant human leptin adsorbed onto aluminum hydroxide gel. | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism | |
3952 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
6 | |
Western Blot, ELISA | |
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction