Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin/OB Antibody (7E10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259497JF549
This item is not returnable.
View return policy
Description
Leptin/OB Monoclonal antibody specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Leptin/OB | |
Monoclonal | |
Janelia Fluor 549 | |
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Recombinant human Leptin/OB (22-167aa) purified from E. coli. | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism | |
3952 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunofluorescence | |
70000000000 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction