Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin/OB Mouse anti-Human, Clone: 11, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1050470.025MG
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.Specifications
Leptin/OB | |
Monoclonal | |
1.0 mg/ml | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
Mouse | |
Protein A or G purified | |
RUO | |
3952 | |
Store at 4°C. Do not freeze. | |
IgG1 κ |
ELISA | |
11 | |
Unconjugated | |
FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Recombinant human leptin adsorbed onto aluminum hydroxide gel | |
0.025 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction