Learn More
Invitrogen™ Leptin Polyclonal Antibody

Description
PA1-052 detects leptin from human, mouse, and porcine samples. PA1-052 has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ~16 kDa protein representing leptin from human serum. The PA1-052 immunogen is a synthetic peptide corresponding to residues S(91) R N V I Q I S N D L E N L R D(106) of human Leptin. This sequence is 94% conserved between human and bovine.
Specifications
Specifications
| Antigen | Leptin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Lep |
| Gene Accession No. | P41159, P41160, Q29406 |
| Gene Alias | CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin |
| Gene Symbols | Lep |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.