Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptin R Antibody (RM0085-9C29), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121725AF700
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.Specifications
Leptin R | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LEPR | |
Mouse recombinant EC-domain of LeptinR | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0085-9C29 | |
Western Blot | |
CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
3953 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction