Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Leptin Receptor Monoclonal Antibody (16)
GREENER_CHOICE

Catalog No. PIMA529739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529739 Supplier Invitrogen™ Supplier No. MA529739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Leptin Receptor/CD295.

The leptin (OB-R) receptor is a member of the cytokine receptor superfamily which plays an important role in mammalian body weight homeostasis and energy balance. Ligand-induced activation of the OB-R by the adipose tissue-secreted hormone, leptin, appears to activate downstream signaling events through the Janus kinase (JAK)/signal transducer and activator of transcription (STAT) pathway. Leptin can homo-dimerize OB-R extracellular domains while downstream signaling events are initiated upon homo-dimerization of OB-R intracellular domains. Several isoforms of the OB-R which differ in length and signaling capabilities have been reported. RNA transcripts encoding the OB-R long form have been found in the choroid plexus and in the hypothalamus which has been proposed as a control center for satiety and energy expenditure. Mutations in either the mouse OB-R (db) or leptin (ob) genes have been shown to result in early-onset obesity. Metabolic abnormalities attributable to these genotypes include hypercorticoidemia, hyperinsulinemia and insulin resistance, hyperglycemia, altered CNS activity, reduced metabolic rate of brown adipose tissue, and a large increase in white adipose tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leptin Receptor
Applications ELISA
Classification Monoclonal
Clone 16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Lepr
Gene Accession No. P48357
Gene Alias B219; CD antigen CD295; CD295; CD295 antigen; db; diabetes; Fa; HuB219; HuB219.2; LEPR; LEP-R; Leprb; LEPRD; LEPROT; leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; Modb1; OB receptor; obese-like; obl; Obr; OB-R; OB-RGRP
Gene Symbols Lepr
Host Species Mouse
Immunogen Recombinant Human Leptin Receptor/CD295 Protein (Met1-Asp839).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.