Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leptospira Biflexa Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $548.00
Specifications
Antigen | Leptospira Biflexa |
---|---|
Concentration | 4 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunofluorescence, Immunocytochemistry, ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017586
![]() |
Novus Biologicals
NB100644830.1ML |
0.1 mL |
Each for $181.90
|
|
|||||
NB10064483
![]() |
Novus Biologicals
NB10064483 |
0.5 mL |
Each for $548.00
|
|
|||||
Description
Leptospira Biflexa Polyclonal specifically detects Leptospira Biflexa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Leptospira Biflexa | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline with 0.1% Sodium Azide | |
IgG | |
IgG purified | |
Reacts with the following Leptospira species: L. biflexa, L. australis, L. canicola, L. grippotyphosa, L. icterohaemorrhagiae, L. pomona and L. serjroe. |
4 mg/mL | |
Immunofluorescence, Immunocytochemistry, ELISA | |
Unconjugated | |
Rabbit | |
Bacteria | |
Native, whole organism | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title